ENA402267095 Enamine

5-(aminomethyl)-2-methylpyrimidin-4-ol

  • MDL number MFCD11108245
  • Cas Number : 1749-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122063
Availability details for
Quantity
  • Availability
  • Add to Cart