ENA402267096 Enamine

2-[(4-tert-butylphenyl)methyl]propanedioic acid

  • MDL number MFCD03003359
  • Cas Number : 683215-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130739
Availability details for
Quantity
  • Availability
  • Add to Cart