ENA402267109 Enamine

tert-butyl N-[3-(3-hydroxypropyl)phenyl]carbamate

  • MDL number MFCD25371569
  • Cas Number : 198896-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122996

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122996
Availability details for
Quantity
  • Availability
  • Add to Cart