ENA402267111 Enamine

4-[4-(aminomethyl)-1-methyl-1H-pyrazol-3-yl]benzonitrile hydrochloride

  • MDL number MFCD25371646
  • Cas Number : 1461707-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130774

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130774
Availability details for
Quantity
  • Availability
  • Add to Cart