ENA402267117 Enamine

methyl 5-sulfamoyl-1,3-thiazole-4-carboxylate

  • MDL number MFCD25371651
  • Cas Number : 89502-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-140
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130786

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130786
Availability details for
Quantity
  • Availability
  • Add to Cart