ENA402267118 Enamine

9-amino-6-oxa-2lambda6-thiaspiro[4.5]decane-2,2-dione hydrochloride

  • MDL number MFCD25371573
  • Cas Number : 1444025-85-7
  • Molecular Weight : 241.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 9-amino-6-oxa-2-thiaspiro[4.5]decane 2,2-dioxide hydrochloride
Inchi Code 1S/C8H15NO3S.ClH/c9-7-1-3-12-8(5-7)2-4-13(10,11)6-8;/h7H,1-6,9H2;1H
Inchi Key YNELMAQQRJSNLW-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-124716

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-124716
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart