ENA402267124 Enamine

methyl 5-(benzylsulfanyl)-1,3-thiazole-4-carboxylate

  • MDL number MFCD20133444
  • Cas Number : 89502-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92...94
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130864

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130864
Availability details for
Quantity
  • Availability
  • Add to Cart