ENA402267128 Enamine

4-chloro-2-(pyridin-2-yl)-1H-1,3-benzodiazole

  • MDL number MFCD20390954
  • Cas Number : 71635-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130870

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130870
Availability details for
Quantity
  • Availability
  • Add to Cart