ENA402267130 Enamine

methyl 4-methoxypentanoate

  • MDL number MFCD25371654
  • Cas Number : 818-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130872
Availability details for
Quantity
  • Availability
  • Add to Cart