ENA402267135 Enamine

(1-{[2-(aminomethyl)phenyl]methyl}pyrrolidin-2-yl)methanol

  • MDL number MFCD14660593
  • Cas Number : 1247814-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130882

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130882
Availability details for
Quantity
  • Availability
  • Add to Cart