ENA402267141 Enamine

tert-butyl 3-hydrazinylpropanoate

  • MDL number MFCD14707427
  • Cas Number : 107349-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130894

Safety Information

signalword Danger
pcodes P201,P202,P210,P261,P264,P270,P271,P280,P281,P301+P312,P302+P352,P304+P340,P305+P351+P338,P308+P313,P310,P312,P330,P363,P370+P378,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-130894
pictograms GHS05,GHS07,GHS08
hstatements H227,H302,H312,H318,H332,H351
Availability details for
Quantity
  • Availability
  • Add to Cart