ENA402267142 Enamine

5-methoxy-1,3-dimethyl-1H-pyrazole-4-carbonitrile

  • MDL number MFCD18879316
  • Cas Number : 1343203-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130895

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130895
Availability details for
Quantity
  • Availability
  • Add to Cart