ENA402267146 Enamine

(1-benzyl-2-methylpiperidin-2-yl)methanol

  • MDL number MFCD25371655
  • Cas Number : 1461706-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130900
Availability details for
Quantity
  • Availability
  • Add to Cart