ENA402267152 Enamine

6-amino-N-methylpyridine-3-sulfonamide

  • MDL number MFCD07440121
  • Cas Number : 94924-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130929

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130929
Availability details for
Quantity
  • Availability
  • Add to Cart