ENA402267157 Enamine

N-(2-hydroxyethyl)bicyclo[2.2.1]hept-5-ene-2-carboxamide

  • MDL number MFCD13678806
  • Cas Number : 75639-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130934
Availability details for
Quantity
  • Availability
  • Add to Cart