ENA402267162 Enamine

2-[2-(propan-2-yl)phenoxy]benzonitrile

  • MDL number MFCD11115711
  • Cas Number : 1036528-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130947

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130947
Availability details for
Quantity
  • Availability
  • Add to Cart