ENA402267166 Enamine

2-bromo-1-chloronaphthalene

  • MDL number MFCD17012571
  • Cas Number : 692728-68-0
  • Molecular Weight : 241.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 56-57
Iupac Name 2-bromo-1-chloronaphthalene
Inchi Code 1S/C10H6BrCl/c11-9-6-5-7-3-1-2-4-8(7)10(9)12/h1-6H
Inchi Key ORPYLLUOBDZSEX-UHFFFAOYSA-N

Description

Melting Point 56-57
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-125103

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-125103
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart