ENA402267167 Enamine

1-(4-fluorophenyl)-1H,4H,5H,6H,7H-imidazo[4,5-c]pyridine dihydrochloride

  • MDL number MFCD25371660
  • Cas Number : 1444317-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130979

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130979
Availability details for
Quantity
  • Availability
  • Add to Cart