ENA402267170 Enamine

2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoic acid hydrate

  • MDL number MFCD11519226
  • Cas Number : 204191-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130994

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130994
Availability details for
Quantity
  • Availability
  • Add to Cart