ENA402267182 Enamine

2-(2-ethyloxan-4-yl)acetic acid

  • MDL number MFCD19651655
  • Cas Number : 1341054-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131026
Availability details for
Quantity
  • Availability
  • Add to Cart