ENA402267183 Enamine

7-bromo-5-methyl-1-benzofuran-2-carboxylic acid

  • MDL number MFCD13187179
  • Cas Number : 35700-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-131027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131027
Availability details for
Quantity
  • Availability
  • Add to Cart