ENA402267194 Enamine

1-[1-(4-bromo-2-methylphenyl)-1H-1,2,3-triazol-4-yl]-2,2,2-trifluoroethan-1-one

  • MDL number MFCD17508158
  • Cas Number : 1461714-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131053
Availability details for
Quantity
  • Availability
  • Add to Cart