ENA402267196 Enamine

5'-bromo-[2,2'-bithiophene]-5-carboxylic acid

  • MDL number MFCD06203205
  • Cas Number : 3339-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131055

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131055
Availability details for
Quantity
  • Availability
  • Add to Cart