ENA402267198 Enamine

7-fluoro-2,3,4,5-tetrahydro-1H-1-benzazepine-2,5-dione

  • MDL number MFCD23097220
  • Cas Number : 476363-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131062

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131062
Availability details for
Quantity
  • Availability
  • Add to Cart