ENA402267201 Enamine

6-(piperidine-1-carbonyl)pyridin-3-amine

  • MDL number MFCD24083042
  • Cas Number : 1301214-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...172
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131071

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131071
Availability details for
Quantity
  • Availability
  • Add to Cart