ENA402267202 Enamine

(5-bromo-1,3-thiazol-2-yl)methanol

  • MDL number MFCD21728921
  • Cas Number : 911052-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73...75
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125440

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125440
Availability details for
Quantity
  • Availability
  • Add to Cart