ENA402267205 Enamine

5-(morpholine-4-sulfonyl)pyridin-2-amine

  • MDL number MFCD06307828
  • Cas Number : 627840-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131077
Availability details for
Quantity
  • Availability
  • Add to Cart