ENA402267211 Enamine

[(imidazolidin-2-ylidene)amino]formonitrile

  • MDL number MFCD01628333
  • Cas Number : 36982-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131087

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131087
Availability details for
Quantity
  • Availability
  • Add to Cart