ENA402267219 Enamine

4-bromo-2-[(2-methanesulfonylethyl)sulfanyl]benzaldehyde

  • MDL number MFCD18887643
  • Cas Number : 1340309-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-131097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131097
Availability details for
Quantity
  • Availability
  • Add to Cart