ENA402267222 Enamine

methyl 5-aminopyrazine-2-carboxylate

  • MDL number MFCD09753440
  • Cas Number : 13924-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230...232
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131101

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-131101
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart