ENA402267226 Enamine

2-ethoxy-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD13175054
  • Cas Number : 706789-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131110
Availability details for
Quantity
  • Availability
  • Add to Cart