ENA402267228 Enamine

6-bromo-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid

  • MDL number MFCD11206335
  • Cas Number : 1094298-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126742

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-126742
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart