ENA402267231 Enamine

6-fluoro-1-(2-fluorophenyl)-1H-1,3-benzodiazol-2-amine hydrobromide

  • MDL number MFCD25371679
  • Cas Number : 1461706-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131117

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131117
Availability details for
Quantity
  • Availability
  • Add to Cart