ENA402267237 Enamine

3,4-dihydro-1H-2-benzopyran-1-carbonitrile

  • MDL number MFCD00457644
  • Cas Number : 13328-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126839

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126839
Availability details for
Quantity
  • Availability
  • Add to Cart