ENA402267238 Enamine

3-(3-fluorophenyl)-1,2,4-oxadiazole-5-thiol

  • MDL number MFCD17259824
  • Cas Number : 1342386-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93...95
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126970

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126970
Availability details for
Quantity
  • Availability
  • Add to Cart