ENA402267241 Enamine

methyl 4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-3-carboxylate

  • MDL number MFCD13179504
  • Cas Number : 132255-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64...66
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131164

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131164
Availability details for
Quantity
  • Availability
  • Add to Cart