ENA402267251 Enamine

bicyclo[2.2.1]heptane-2-carbonitrile

  • Cas Number : 2234-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43-45
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131202
Availability details for
Quantity
  • Availability
  • Add to Cart