ENA402267252 Enamine

6-methyl-1H,6H,7H-pyrrolo[2,3-c]pyridin-7-one

  • MDL number MFCD12924072
  • Cas Number : 116212-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131203

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131203
Availability details for
Quantity
  • Availability
  • Add to Cart