ENA402267255 Enamine

2-(3-amino-1H-pyrazol-1-yl)-5-chlorobenzonitrile

  • MDL number MFCD16769639
  • Cas Number : 1249223-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131206

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131206
Availability details for
Quantity
  • Availability
  • Add to Cart