ENA402267268 Enamine

methyl 2-(3-methyl-4-nitro-1H-pyrazol-1-yl)acetate

  • MDL number MFCD04967344
  • Cas Number : 1005640-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-131223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131223
Availability details for
Quantity
  • Availability
  • Add to Cart