ENA402267269 Enamine

4-chloro-3-methyl-2-(trifluoromethyl)quinoline

  • MDL number MFCD18876619
  • Cas Number : 1344242-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131224
Availability details for
Quantity
  • Availability
  • Add to Cart