ENA402267276 Enamine

3-chloro-N-ethyl-2-methylaniline

  • MDL number MFCD03210691
  • Cas Number : 75746-96-2
  • Molecular Weight : 169.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-131237

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131237
Availability details for
Quantity
  • Availability
  • Add to Cart