ENA402267278 Enamine

3,5-dimethoxycyclohexan-1-ol

  • MDL number MFCD25371584
  • Cas Number : 191865-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127346

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127346
Availability details for
Quantity
  • Availability
  • Add to Cart