ENA402267284 Enamine

2-amino-2-(3,4-dimethoxyphenyl)propanoic acid

  • MDL number MFCD02662564
  • Cas Number : 4454-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101744

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101744
Availability details for
Quantity
  • Availability
  • Add to Cart