ENA402267292 Enamine

4-cyclopropylpiperidine hydrochloride

  • MDL number MFCD22573739
  • Cas Number : 208245-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 2102-2103
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131343

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131343
Availability details for
Quantity
  • Availability
  • Add to Cart