ENA402267294 Enamine

1-bromo-2-methoxypropane

  • MDL number MFCD20621723
  • Cas Number : 23465-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131348

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-131348
pictograms GHS02,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart