ENA402267295 Enamine

2-ethyl-5-methyl-1H-1,3-benzodiazole

  • MDL number MFCD06312169
  • Molecular Weight : 160.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-ethyl-5-methyl-1H-benzimidazole
Inchi Code 1S/C10H12N2/c1-3-10-11-8-5-4-7(2)6-9(8)12-10/h4-6H,3H2,1-2H3,(H,11,12)
Inchi Key ICWSZURISQHRLN-UHFFFAOYSA-N

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart