ENA402267305 Enamine

4-(3-amino-1H-pyrazol-1-yl)-3-fluorobenzonitrile

  • MDL number MFCD14594759
  • Cas Number : 1249090-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131363

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131363
Availability details for
Quantity
  • Availability
  • Add to Cart