ENA402267310 Enamine

[(4-methylimidazolidin-2-ylidene)amino]formonitrile

  • Cas Number : 49552-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131371
Availability details for
Quantity
  • Availability
  • Add to Cart