ENA402267312 Enamine

2-chloro-4-[3-(propan-2-yl)-1,2,4-oxadiazol-5-yl]aniline

  • MDL number MFCD20296386
  • Cas Number : 1343684-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131373

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131373
Availability details for
Quantity
  • Availability
  • Add to Cart